missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Imject™ Freund's Adjuvants

Stimulate a nonspecific immune response using complete (CFA) or incomplete (IFA) formulations of the popular Freund's adjuvant; offered in 10mL ampules.

$132.60 - $146.88

Specifications

Quantity 5 x 10mL
Sufficient For Mixture with 50mL of immunogen
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI77140 Thermo Scientific™
PI77140
Imject Freund's Complete Adjuvant (FCA) Pack of 5 Each for $146.88
PI77145 Thermo Scientific™
PI77145
Imject Freund's Incomplete Adjuvant (FIA) Pack of 5 Each for $132.60
Specifications

Description

Thermo Scientific™ Imject Freund's Adjuvants are 10mL ampules of complete (FCA) and incomplete (FIA) formulations of a popular nonspecific stimulator of the immune response for use in antibody production immunization protocols.

The water-and-oil emulsions (with and without inactivated Mycobacterium) greatly enhance immune responses to immunogens when mixed and injected with the prepared antigen. Typically, FCA is used for initial injections and FIA is used for booster injections. The ampules comprise conveniently sized aliquots that provide long shelf life.

Highlights:

  • Water-in-oil emulsion with or without killed Mycobacterium
  • Enhance immune response and antibody production for injected immunogens
  • Useful for immunization of mice, rabbits and other mammalian hosts
  • Requires emulsification with prepared immunogens

Freund's Complete Adjuvant (FCA), Part No. 77140:

  • Water-in-oil emulsion containing killed Mycobacterium
  • Greatly enhances immune response to immunogen
  • Typically used for initial injections
  • Disadvantages: difficult to mix with immunogen and can cause tissue necrosis at injection site

Freund's Incomplete Adjuvant (FIA), Part No. 77145:

  • Water-in-oil emulsion without Mycobacterium
  • Enhances immune response in booster injections
  • Typically used for subsequent boosts after initial injection with FCA
  • Disadvantage: difficult to mix with immunogen
Description & Specifications

Specifications

5 x 10mL
Mixture with 50mL of immunogen