missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iminodiacetic Acid 98.0+%, TCI America™

$28.21 - $76.32

Chemical Identifiers

CAS 142-73-4
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.103
MDL Number MFCD00004280
InChI Key NBZBKCUXIYYUSX-UHFFFAOYSA-N
Synonym iminodiacetic acid, 2,2'-iminodiacetic acid, glycine, n-carboxymethyl, aminodiacetic acid, diglycocoll, diglykokoll, diglycin, iminodiethanoic acid, n-carboxymethyl glycine, 2,2'-azanediyldiacetic acid
PubChem CID 8897
ChEBI CHEBI:24786
IUPAC Name 2-(carboxymethylamino)acetic acid
SMILES C(C(=O)O)NCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I000625G
View Documents Promotion Details
TCI America
I000625G
25 g
Each for $28.21
 
I0006500G
View Documents Promotion Details
TCI America
I0006500G
500 g
Each for $76.32
 
Description

Description

Specifications

Chemical Identifiers

142-73-4
133.103
NBZBKCUXIYYUSX-UHFFFAOYSA-N
8897
2-(carboxymethylamino)acetic acid
C4H7NO4
MFCD00004280
iminodiacetic acid, 2,2'-iminodiacetic acid, glycine, n-carboxymethyl, aminodiacetic acid, diglycocoll, diglykokoll, diglycin, iminodiethanoic acid, n-carboxymethyl glycine, 2,2'-azanediyldiacetic acid
CHEBI:24786
C(C(=O)O)NCC(=O)O

Specifications

142-73-4
White
MFCD00004280
iminodiacetic acid, 2,2'-iminodiacetic acid, glycine, n-carboxymethyl, aminodiacetic acid, diglycocoll, diglykokoll, diglycin, iminodiethanoic acid, n-carboxymethyl glycine, 2,2'-azanediyldiacetic acid
C(C(=O)O)NCC(=O)O
133.103
CHEBI:24786
≥98.0% (T)
Iminodiacetic Acid
250°C
C4H7NO4
25 g
NBZBKCUXIYYUSX-UHFFFAOYSA-N
2-(carboxymethylamino)acetic acid
8897
133.10
Crystalline Powder
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1211

RTECSNumber : AI2975000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'