Please login to your online account to display your discounted pricing

Iminodiacetic Acid, Disodium Salt Monohydrate Pract., 80%, Acros Organics


Quantity 500g

Manufacturer: thermo fisher scientific  412285000

 View more versions of this product

Catalog No. AC412285000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥75% (on dry substance)
Physical Form Powder
Formula Weight 195.08
Quantity 500g
Color White to slightly brown
Merck Index 10, 4815
Beilstein 04, 365
Synonym Disodium iminodiacetate monohydrate
Chemical Name or Material Iminodiacetic acid, disodium salt monohydrate
Infrared Spectrum Authentic
Name Note pract., 80%
Loss on Drying 20% max. (105°C)
CAS 17593-73-6
Molecular Formula C4H5NNa2O4·H2O
Linear Formula HN(CH2CO2Na)2·H2O
Assay Percent Range 80%

Specifications

Chemical Name or Material Iminodiacetic acid, disodium salt monohydrate
Name Note pract., 80%
CAS 17593-73-6
Assay ≥75% (on dry substance)
Assay Percent Range 80%
Molecular Formula C4H5NNa2O4·H2O
Linear Formula HN(CH2CO2Na)2·H2O
Formula Weight 195.08
Synonym Disodium iminodiacetate monohydrate