Please login to your online account to display your discounted pricing

Iminodiacetic Acid, Disodium Salt Monohydrate Pract., 80%, Acros Organics

$188.09 - $188.09

Chemical Identifiers

Chemical Name or Material Iminodiacetic acid, disodium salt monohydrate
Name Note pract., 80%
CAS 17593-73-6
Assay ≥75% (on dry substance)
Assay Percent Range 80%
Molecular Formula C4H5NNa2O4·H2O
Linear Formula HN(CH2CO2Na)2·H2O
Formula Weight 195.08
Synonym Disodium iminodiacetate monohydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC412285000

  SDS
thermo fisher scientific
412285000
500g Each for $188.09
Description & Specifications

Specifications

Assay ≥75% (on dry substance)
Physical Form Powder
Formula Weight 195.08
Quantity 500g
Color White to slightly brown
Beilstein 04, 365
Merck Index 10, 4815
Chemical Name or Material Iminodiacetic acid, disodium salt monohydrate
Infrared Spectrum Authentic
Loss on Drying 20% max. (105°C)
Name Note pract., 80%
Assay Percent Range 80%
Molecular Formula C4H5NNa2O4·H2O
Linear Formula HN(CH2CO2Na)2·H2O
CAS 17593-73-6
Synonym Disodium iminodiacetate monohydrate