Please login to your online account to display your discounted pricing

Iminodiacetic acid, 98+%, ACROS Organics™


Quantity 2.5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC204970025

 View more versions of this product

Catalog No. AC204970025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Crystalline Powder
Formula Weight 133.1
Quantity 2.5kg
Color White or yellow
TSCA TSCA
RTECS Number AI2975000
EINECS Number 205-555-4
Beilstein 04, 365
Merck Index 15, 4959
Chemical Name or Material Iminodiacetic acid
Infrared Spectrum Authentic
Packaging Plastic bottle
Name Note 98+%
Assay Percent Range 98+%
Molecular Formula C4H7NO4
Linear Formula HN(CH2CO2H)2
MDL Number MFCD00004280
CAS 142-73-4
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Iminodiacetic acid
Name Note 98+%
CAS 142-73-4
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C4H7NO4
Linear Formula HN(CH2CO2H)2
Formula Weight 133.1
MDL Number MFCD00004280
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents