missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iminodiacetic acid, 98+%

Manufacturer:  Alfa Aesar™ A1151022

 View more versions of this product

Catalog No. AAA1151022


 

Description

Description

Applications
Iminoacetic acid is used as an inhibitor of bovine liver glutamate dehydrogenase. It is used as a reagent in the synthesis of poly(N-isopropylacrylamide) (PNIPAM) microgel particles for metal affinity binding of peptides. It is mainly used for modulating peptide mobility by capillary electrophoresis. It is also used in chemotherapeutic treatment for metal poisoning. It acts as a tridendate ligand and forms metal complexes.

Solubility
Soluble in water. Insoluble in ethanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Specifications

Iminodiacetic acid
≥98%
MFCD00004280
14,4917
C(C(=O)O)NCC(=O)O
~243°C (decomposition)
8897
133.1
142-73-4
C4H7NO4
878499
NBZBKCUXIYYUSX-UHFFFAOYSA-N
2-(carboxymethylamino)acetic acid
133.103
CHEBI:24786
100g

Chemical Identifiers

142-73-4
133.103
NBZBKCUXIYYUSX-UHFFFAOYSA-N
CHEBI:24786
C4H7NO4
MFCD00004280
8897
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AI2975000

TSCA : TSCA

SDS Documents