missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Imidazole 98.0+%, TCI America™
$28.21 - $131.06
Chemical Identifiers
CAS | 288-32-4 |
---|---|
Molecular Formula | C3H4N2 |
Molecular Weight (g/mol) | 68.08 |
MDL Number | MFCD00005183 |
InChI Key | RAXXELZNTBOGNW-UHFFFAOYSA-N |
Synonym | imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole |
PubChem CID | 795 |
ChEBI | CHEBI:16069 |
IUPAC Name | 1H-imidazole |
SMILES | N1C=CN=C1 |
Chemical Identifiers
288-32-4 | |
68.08 | |
RAXXELZNTBOGNW-UHFFFAOYSA-N | |
795 | |
1H-imidazole |
C3H4N2 | |
MFCD00005183 | |
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole | |
CHEBI:16069 | |
N1C=CN=C1 |
Specifications
288-32-4 | |
White | |
C3H4N2 | |
25 g | |
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole | |
N1C=CN=C1 | |
68.08 | |
CHEBI:16069 | |
≥98.0% (T) | |
Imidazole |
90°C | |
256°C | |
MFCD00005183 | |
2923 | |
RAXXELZNTBOGNW-UHFFFAOYSA-N | |
1H-imidazole | |
795 | |
68.08 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-0381
RTECSNumber : NI3325000
TSCA : Yes