missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Imidazole, sodium derivative, ACROS Organics™

Manufacturer:  Acros Organics AC188980050

 View more versions of this product

Catalog No. AC188980050



Specifications

Specifications

Imidazole, sodium derivative
3% max.
Orange to Yellow
Sodium derivative
C3H3N2Na
23, II, 35
ITAWMPSVROAMOE-UHFFFAOYSA-N
sodium;imidazol-3-ide
4605971
Free Flowing Crystalline Powder
Authentic
Glass bottle
5g
5587-42-8
MFCD00064314
sodium imidazolide, imidazole sodium derivative, imidazolylsodium, sodium imidazolate, imidazol-1-yl sodium, sodium imidazol-3-ide, sodium imidazol-1-ide, 1-sodio-1h-imidazole, 1-sodioimidazole, imidazole sodium derivative, technical grade
C1=CN=C[N-]1.[Na+]
90.061
90.06
≥97.0%

Chemical Identifiers

5587-42-8
90.061
ITAWMPSVROAMOE-UHFFFAOYSA-N
4605971
C1=CN=C[N-]1.[Na+]
C3H3N2Na
MFCD00064314
sodium imidazolide, imidazole sodium derivative, imidazolylsodium, sodium imidazolate, imidazol-1-yl sodium, sodium imidazol-3-ide, sodium imidazol-1-ide, 1-sodio-1h-imidazole, 1-sodioimidazole, imidazole sodium derivative, technical grade
sodium;imidazol-3-ide
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents