Please login to your online account to display your discounted pricing

Alfa Aesar™ Imidazole, Molecular Biology Grade, ≥99%


Quantity 25g

Manufacturer: alfa aesar™  J6713214

 View more versions of this product

Catalog No. AAJ6713214

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 68.08
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Imidazole
Health Hazard 1 Danger
UN Number UN2923
MDL Number MFCD00005183
Assay ≥99%
TSCA TSCA
Health Hazard 2 GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Purity Grade Molecular Biology Grade
Physical Form Powder
Quantity 25g
Melting Point 90°C
Merck Index 144,912
CAS 288-32-4
Synonym Glyoxaline
Beilstein 103853
Boiling Point 256°C
DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
RTECS Number NI3325000
Color White to off-white
Density 1.03g/mL

Specifications

Chemical Name or Material Imidazole
CAS 288-32-4
Purity Grade Molecular Biology Grade
Formula Weight 68.08
MDL Number MFCD00005183
UN Number UN2923
Beilstein 103853
Merck Index 144,912
Synonym Glyoxaline
Accessories

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

SDS & Safety Documents