missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Imidazole 99.0+%, TCI America™

$167.88 - $2,281.19

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.079
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
Synonym 1,3-diaza-2,4-cyclopentadiene
PubChem CID 795
ChEBI CHEBI:16069
IUPAC Name 1H-imidazole
SMILES C1=CN=CN1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I028825G View Documents Tci America
I028825G
25g Each for $167.88
 
I0288100G View Documents Tci America
I0288100G
100g Each for $669.88
 
I0288500G View Documents Tci America
I0288500G
500g Each for $2,281.19
 
Specifications

Chemical Identifiers

288-32-4
68.079
RAXXELZNTBOGNW-UHFFFAOYSA-N
795
1H-imidazole
C3H4N2
MFCD00005183
1,3-diaza-2,4-cyclopentadiene
CHEBI:16069
C1=CN=CN1

Specifications

256°C
90°C
288-32-4
MFCD00005183
1,3-diaza-2,4-cyclopentadiene
C1=CN=CN1
68.079
CHEBI:16069
Crystal-Powder at 20°C
Imidazole [for Buffer]
White
25g
C3H4N2
2923
RAXXELZNTBOGNW-UHFFFAOYSA-N
1H-imidazole
795
68.08
≥99.0% (GC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0381

RTECSNumber : NI3325000

TSCA : Yes