missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Imidazole-buffered saline (5X)

$90.36 - $128.31

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.079
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
PubChem CID 795
ChEBI CHEBI:16069
IUPAC Name 1H-imidazole
SMILES C1=CN=CN1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63887AK View Documents Alfa Aesar™
J63887AK
250mL Each for $90.36
 
AAJ63887AP View Documents Alfa Aesar™
J63887AP
500mL Each for $128.31
 
Specifications

Chemical Identifiers

288-32-4
68.079
RAXXELZNTBOGNW-UHFFFAOYSA-N
CHEBI:16069
C1=CN=CN1
C3H4N2
MFCD00005183
795
1H-imidazole

Specifications

288-32-4
MFCD00005183
RAXXELZNTBOGNW-UHFFFAOYSA-N
1H-imidazole
795
Liquid
250mL
C3H4N2
5X
C1=CN=CN1
68.079
CHEBI:16069
Odorless
Imidazole-buffered saline
Safety and Handling

Safety and Handling

TSCA : TSCA