Learn More
Imidazole, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 396745000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Imidazole | |
288-32-4 | |
White to Yellow | |
255.0°C to 256.0°C | |
0.1% max. | |
C3H4N2 | |
500 g | |
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole | |
RAXXELZNTBOGNW-UHFFFAOYSA-N | |
1H-imidazole | |
795 | |
68.08 | |
ACS Reagent |
For analysis ACS | |
86.0°C to 90.0°C | |
9.5 to 11.0 (at 5% 25°C) | |
145°C | |
Plastic bottle | |
MFCD00005183 | |
15, 4953 | |
Solubility in water: 633g/L (20°C). Other solubilities: freely soluble in alcohol,ether,chloroform,slightly soluble in benzene,sparingly soluble in petroleum ether | |
N1C=CN=C1 | |
68.08 | |
CHEBI:16069 | |
≥99% | |
Crystals or Powder |
Chemical Identifiers
288-32-4 | |
68.08 | |
RAXXELZNTBOGNW-UHFFFAOYSA-N | |
795 | |
1H-imidazole |
C3H4N2 | |
MFCD00005183 | |
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole | |
CHEBI:16069 | |
N1C=CN=C1 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May damage the unborn child.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
GHS Signal Word: Danger
EINECSNumber : 206-019-2
RUO – Research Use Only