Learn More
Imidazole, 99+%, crystalline, Thermo Scientific Chemicals
Imidazole, 99+%, C3H4N2, CAS Number-288-32-4, glyoxalin, imidazole, miazole, imutex, pyrro b monazole, 1,3-diazole, imidazol, iminazole, 1,3-diaza-2,4-cyclopentadiene, glyoxaline, 1kg, 99% min. (GC), 23, 45, 255.0 deg.C to 256.0 deg.C, CHEBI:16069, White to Yellow or Beige, 206-019-2, 01,492; 02,220
$138.93 - $683.88
Chemical Identifiers
CAS | 288-32-4 |
---|---|
Molecular Formula | C3H4N2 |
Molecular Weight (g/mol) | 68.08 |
MDL Number | MFCD00005183 |
InChI Key | RAXXELZNTBOGNW-UHFFFAOYSA-N |
Synonym | imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole |
PubChem CID | 795 |
ChEBI | CHEBI:16069 |
IUPAC Name | 1H-imidazole |
SMILES | N1C=CN=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC301872500
|
Thermo Scientific Chemicals
301872500 |
250 g | Plastic bottle |
Each for $138.93
|
|
||||
AC301870010
|
Thermo Scientific Chemicals
301870010 |
1 kg | Plastic bottle |
Each for $421.21
|
|
||||
AC301870025
|
Thermo Scientific Chemicals
301870025 |
2.5 kg | Plastic bottle |
Each for $683.88
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
288-32-4 | |
68.08 | |
RAXXELZNTBOGNW-UHFFFAOYSA-N | |
795 | |
1H-imidazole |
C3H4N2 | |
MFCD00005183 | |
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole | |
CHEBI:16069 | |
N1C=CN=C1 |
Specifications
288-32-4 | |
White to Yellow or Beige | |
145°C | |
99% min. (GC) | |
C3H4N2 | |
250 g | |
01,492; 02,220 | |
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole | |
RAXXELZNTBOGNW-UHFFFAOYSA-N | |
1H-imidazole | |
795 | |
68.08 | |
Crystalline Powder |
86.0°C to 90.0°C | |
255.0°C to 256.0°C | |
Authentic | |
Plastic bottle | |
MFCD00005183 | |
23, 45 | |
15, 4953 | |
Solubility in water: 633g/L (20°C). Other solubilities: freely soluble in alcohol,ether,chloroform,slightly soluble in benzene,sparingly soluble in petroleum ether | |
N1C=CN=C1 | |
68.08 | |
CHEBI:16069 | |
99+% | |
Imidazole |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May damage the unborn child.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
GHS Signal Word: Danger
EINECSNumber : 206-019-2
RUO – Research Use Only