Learn More
Imidazole, 99%, Thermo Scientific Chemicals
Imidazole, >-98.5%, C3H4N2, CAS Number-288-32-4, 1,3-diaza-2,4-cyclopentadiene, 1,3-diazole, glyoxalin, glyoxaline, imidazol, imidazole, iminazole, imutex, miazole, pyrro b monazole, 500g, 99%, 23, 45, 255.0 deg.C to 256.0 deg.C, CHEBI:16069, White to Yellow, 01,492; 02,220, 145 deg.C, 68.08
Supplier: Thermo Scientific Chemicals 122025000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Imidazole | |
86.0°C to 90.0°C | |
255.0°C to 256.0°C | |
Authentic | |
Plastic Bottle | |
MFCD00005183 | |
23, 45 | |
15, 4953 | |
RAXXELZNTBOGNW-UHFFFAOYSA-N | |
1H-imidazole | |
795 | |
68.08 | |
≥98.5% |
288-32-4 | |
White to Yellow | |
145°C | |
99% | |
C3H4N2 | |
500 g | |
01,492; 02,220 | |
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole | |
N1C=CN=C1 | |
68.08 | |
CHEBI:16069 | |
0.003mbar at 20°C | |
Flakes |
Chemical Identifiers
288-32-4 | |
68.08 | |
RAXXELZNTBOGNW-UHFFFAOYSA-N | |
795 | |
1H-imidazole |
C3H4N2 | |
MFCD00005183 | |
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole | |
CHEBI:16069 | |
N1C=CN=C1 |
Safety and Handling
Emergency Overview
Causes burns by all exposure routes. Harmful if swallowed. Possible risk of harm to the unborn child.
NFPA
Health:3 Flammability:1 Instability:0
Signal Word: DANGER!
RUO – Research Use Only