Please login to your online account to display your discounted pricing

Imidazole, >99.6%, MP Biomedicals™

$1,073.82 - $1,073.82

Chemical Identifiers

Chemical Name or Material Imidazole
CAS 288-32-4
Physical Form Powder
Assay Percent Range >99.6%
Molecular Formula C3H4N2
Formula Weight 68.08
Synonym 1, 3-Diazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN10203305

 
mp biomedicals
10203305
5kg Each for $1,073.82
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 68.08
Color White
Quantity 5kg
Density 500 - 600kg/m3 (bulk density)
Molecular Formula C3H4N2
Chemical Name or Material Imidazole
Assay Percent Range >99.6%
CAS 288-32-4
Synonym 1, 3-Diazole