missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ Imidazole-4-carboxylic acid, 98%
$32.39 - $312.91
Chemical Identifiers
CAS | 1072-84-0 |
---|---|
Molecular Formula | C4H4N2O2 |
Molecular Weight (g/mol) | 112.088 |
MDL Number | MFCD00082203 |
InChI Key | NKWCGTOZTHZDHB-UHFFFAOYSA-N |
Synonym | 1h-imidazole-4-carboxylic acid, 3h-imidazole-4-carboxylic acid, 4 5-imidazolecarboxylic acid, 4-carboxyimidazole, 4-imidazole carboxylic acid, 4-imidazolecarboxylic acid, ae82z8u4gj, imidazole-4-carboxylic acid, imidazole-5-carboxylic acid, unii-ae82z8u4gj |
PubChem CID | 14080 |
IUPAC Name | 1H-imidazole-5-carboxylic acid |
SMILES | C1=C(NC=N1)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH56862MD
|
Alfa Aesar™
H56862MD |
250mg |
Each for $32.39
|
|
AAH5686203
|
Alfa Aesar™
H5686203 |
1g |
Each for $89.74
|
|
AAH5686206
|
Alfa Aesar™
H5686206 |
5g |
Each for $312.91
|
|
Chemical Identifiers
1072-84-0 | |
112.088 | |
NKWCGTOZTHZDHB-UHFFFAOYSA-N | |
14080 | |
C1=C(NC=N1)C(=O)O |
C4H4N2O2 | |
MFCD00082203 | |
1h-imidazole-4-carboxylic acid, 3h-imidazole-4-carboxylic acid, 4 5-imidazolecarboxylic acid, 4-carboxyimidazole, 4-imidazole carboxylic acid, 4-imidazolecarboxylic acid, ae82z8u4gj, imidazole-4-carboxylic acid, imidazole-5-carboxylic acid, unii-ae82z8u4gj | |
1H-imidazole-5-carboxylic acid |
Specifications
294°C to 295°C | |
1072-84-0 | |
MFCD00082203 | |
NKWCGTOZTHZDHB-UHFFFAOYSA-N | |
1H-imidazole-5-carboxylic acid | |
14080 | |
98% |
250mg | |
C4H4N2O2 | |
1h-imidazole-4-carboxylic acid, 3h-imidazole-4-carboxylic acid, 4 5-imidazolecarboxylic acid, 4-carboxyimidazole, 4-imidazole carboxylic acid, 4-imidazolecarboxylic acid, ae82z8u4gj, imidazole-4-carboxylic acid, imidazole-5-carboxylic acid, unii-ae82z8u4gj | |
C1=C(NC=N1)C(=O)O | |
112.088 | |
112.09 | |
Imidazole-4-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-017-8
RTECSNumber : NI4001000
TSCA : No
Recommended Storage : Ambient temperatures