Please login to your online account to display your discounted pricing

Alfa Aesar™ Imidazole-4-carboxaldehyde, 97%

$46.26 - $623.80

Chemical Identifiers

Chemical Name or Material Imidazole-4-carboxaldehyde
CAS 3034-50-2
Molecular Formula C4H4N2O
Formula Weight 96.09
MDL Number MFCD00173726
Synonym 4-Formylimidazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2505003

  SDS
alfa aesar™
B2505003
1g Each for $46.26

AAB2505006

  SDS
alfa aesar™
B2505006
5g Each for $150.68

AAB2505014

  SDS
alfa aesar™
B2505014
25g Each for $623.80
Description & Specifications

Specifications

Formula Weight 96.09
Melting Point 174° to 177°C
Quantity 1g
Assay 97%
Health Hazard 1 Warning
MDL Number MFCD00173726
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 3034-50-2
Chemical Name or Material Imidazole-4-carboxaldehyde
Molecular Formula C4H4N2O
Synonym 4-Formylimidazole
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
Imidazole-4-carboxaldehyde is a 4-formyl derivative of imidazole used in the preparation of C17,20-lyase inhibitor for the treatment of androgen-dependent prostate cancer. It is also used in the synthesis of other biologically active compounds such as antimalarial drugs, fabrication of colorimetric chemosensor.

Solubility
Soluble in DMSO, methanol.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.