Learn More
Imidazole-4-carboxaldehyde, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2505006
Description
Imidazole-4-carboxaldehyde is a 4-formyl derivative of imidazole used in the preparation of C17,20-lyase inhibitor for the treatment of androgen-dependent prostate cancer. It is also used in the synthesis of other biologically active compounds such as antimalarial drugs, fabrication of colorimetric chemosensor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsImidazole-4-carboxaldehyde is a 4-formyl derivative of imidazole used in the preparation of C17,20-lyase inhibitor for the treatment of androgen-dependent prostate cancer. It is also used in the synthesis of other biologically active compounds such as antimalarial drugs, fabrication of colorimetric chemosensor.
Solubility
Soluble in DMSO, methanol.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
Specifications
Imidazole-4-carboxaldehyde | |
174°C to 177°C | |
MFCD00173726 | |
Air Sensitive | |
Soluble in DMSO,methanol. | |
C1=C(NC=N1)C=O | |
96.089 | |
96.09 |
3034-50-2 | |
C4H4N2O | |
5 g | |
1h-imidazole-4-carbaldehyde, imidazole-4-carbaldehyde, 1h-imidazole-4-carboxaldehyde, imidazole-4-carboxaldehyde, 4-imidazolecarboxaldehyde, 4-formylimidazole, 4 5-imidazolecarboxaldehyde, 3h-imidazole-4-carbaldehyde, 4-formyl-1h-imidazole, imidazole-5-carbaldehyde | |
ZQEXIXXJFSQPNA-UHFFFAOYSA-N | |
1H-imidazole-5-carbaldehyde | |
76428 | |
97% |
Chemical Identifiers
3034-50-2 | |
96.089 | |
ZQEXIXXJFSQPNA-UHFFFAOYSA-N | |
76428 | |
C1=C(NC=N1)C=O |
C4H4N2O | |
MFCD00173726 | |
1h-imidazole-4-carbaldehyde, imidazole-4-carbaldehyde, 1h-imidazole-4-carboxaldehyde, imidazole-4-carboxaldehyde, 4-imidazolecarboxaldehyde, 4-formylimidazole, 4 5-imidazolecarboxaldehyde, 3h-imidazole-4-carbaldehyde, 4-formyl-1h-imidazole, imidazole-5-carbaldehyde | |
1H-imidazole-5-carbaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 221-227-3
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only