Please login to your online account to display your discounted pricing

Imidazole-2-carboxaldehyde, 98%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC398470010

 View more versions of this product

Catalog No. AC398470010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 96.09
Quantity 1g
Molecular Formula C4H4N2O
CAS 10111-08-7
Chemical Name or Material Imidazole-2-carboxaldehyde
Assay Percent Range 98%
MDL Number MFCD00003544

Specifications

Chemical Name or Material Imidazole-2-carboxaldehyde
Assay Percent Range 98%
Molecular Formula C4H4N2O
Formula Weight 96.09
MDL Number MFCD00003544