Please login to your online account to display your discounted pricing

Imidazole-2-carboxaldehyde, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Imidazole-2-carboxaldehyde
Assay Percent Range 98%
Molecular Formula C4H4N2O
Formula Weight 96.09
MDL Number MFCD00003544
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC398470010

 
thermo fisher scientific
AC398470010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC398470050

 
thermo fisher scientific
AC398470050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Imidazole-2-carboxaldehyde
CAS 10111-08-7
Assay Percent Range 98%
Molecular Formula C4H4N2O
Formula Weight 96.09
MDL Number MFCD00003544
Quantity 1g