missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Imidazole, 0.5M buffer soln., pH 6.0

$54.59 - $85.27

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.079
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
PubChem CID 795
ChEBI CHEBI:16069
IUPAC Name 1H-imidazole
SMILES C1=CN=CN1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61956AE View Documents Alfa Aesar™
J61956AE
100mL Each for $54.59
 
AAJ61956AK View Documents Alfa Aesar™
J61956AK
250mL Each for $85.27
 
Specifications

Chemical Identifiers

288-32-4
68.079
RAXXELZNTBOGNW-UHFFFAOYSA-N
CHEBI:16069
C1=CN=CN1
C3H4N2
MFCD00005183
795
1H-imidazole

Specifications

288-32-4
MFCD00005183
RAXXELZNTBOGNW-UHFFFAOYSA-N
1H-imidazole
795
Liquid
Colorless
Imidazole buffer soln.
C3H4N2
0.5M buffer soln.
C1=CN=CN1
68.079
CHEBI:16069
6.0
100mL
Safety and Handling

Safety and Handling

GHS H Statement
H318-H315
Causes serious eye damage.
Causes skin irritation.

GHS P Statement
P280-P305+P351+P338-P310-P362-P321-P332+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.

Danger

TSCA : TSCA

Recommended Storage : 2° to 8°C