Please login to your online account to display your discounted pricing

Alfa Aesar™ Imidazole, 0.2M buffer soln., pH 6.0, low endotoxin

Chemical Identifiers

Chemical Name or Material Imidazole
CAS 288-32-4
Name Note low endotoxin
MDL Number MFCD00005183
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67255AE

 
alfa aesar™
J67255AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67255AK

 
alfa aesar™
J67255AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Imidazole
Physical Form Liquid
MDL Number MFCD00005183
CAS 288-32-4
Color Colorless
Name Note low endotoxin
Storage Note 1 2° to 8°C
pH 6.0
Quantity 100mL
TSCA TSCA
Concentration 0.2M buffer soln.
Safety and Handling

2° to 8°C