Please login to your online account to display your discounted pricing

Imidazo[2,1-b]thiazole-6-carbaldehyde 97+%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC32504CB

 View more versions of this product

Catalog No. CC32504CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 152.18
Quantity 250mg
Assay 97%
Chemical Name or Material Imidazo[2, 1-b]thiazole-6-carbaldehyde
Assay Percent Range 97%
Molecular Formula C6H4N2OS
Packaging Amber Glass Bottle
CAS 120107-61-1

Specifications

Chemical Name or Material Imidazo[2, 1-b]thiazole-6-carbaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H4N2OS
Formula Weight 152.18