missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Imidazo[2,1-b]thiazole-6-carbaldehyde 97+%, Thermo Scientific™
Chemical Identifiers
CAS |
120107-61-1 |
Molecular Formula |
C6H4N2OS |
Molecular Weight (g/mol) |
152.171 |
InChI Key |
VYBDPAAWKNKEKY-UHFFFAOYSA-N |
Synonym |
imidazo 2,1-b thiazole-6-carbaldehyde, imidazo 2,1-b 1,3 thiazole-6-carbaldehyde, imidazo 2,1-b thiazole-6-carboxaldehyde, acmc-20moqu, imidazo 2,1-b 1,3-thiazoline-6-carbaldehyde, imidazo 2,1-b 1,3 thiazole-6-carboxaldehyde, imidazo 2,1-b thiazole-6-carboxaldehydeimidazo |
PubChem CID |
4961258 |
IUPAC Name |
imidazo[2,1-b][1,3]thiazole-6-carbaldehyde |
SMILES |
C1=CSC2=NC(=CN21)C=O |
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
|
120107-61-1 |
|
152.171 |
|
imidazo 2,1-b thiazole-6-carbaldehyde, imidazo 2,1-b 1,3 thiazole-6-carbaldehyde, imidazo 2,1-b thiazole-6-carboxaldehyde, acmc-20moqu, imidazo 2,1-b 1,3-thiazoline-6-carbaldehyde, imidazo 2,1-b 1,3 thiazole-6-carboxaldehyde, imidazo 2,1-b thiazole-6-carboxaldehydeimidazo |
|
imidazo[2,1-b][1,3]thiazole-6-carbaldehyde |
|
C6H4N2OS |
|
VYBDPAAWKNKEKY-UHFFFAOYSA-N |
|
4961258 |
|
C1=CSC2=NC(=CN21)C=O |
Specifications
|
120107-61-1 |
|
Amber Glass Bottle |
|
250mg |
|
VYBDPAAWKNKEKY-UHFFFAOYSA-N |
|
imidazo[2,1-b][1,3]thiazole-6-carbaldehyde |
|
4961258 |
|
97% |
|
97% |
|
C6H4N2OS |
|
imidazo 2,1-b thiazole-6-carbaldehyde, imidazo 2,1-b 1,3 thiazole-6-carbaldehyde, imidazo 2,1-b thiazole-6-carboxaldehyde, acmc-20moqu, imidazo 2,1-b 1,3-thiazoline-6-carbaldehyde, imidazo 2,1-b 1,3 thiazole-6-carboxaldehyde, imidazo 2,1-b thiazole-6-carboxaldehydeimidazo |
|
C1=CSC2=NC(=CN21)C=O |
|
152.171 |
|
152.18 |
|
Imidazo[2, 1-b]thiazole-6-carbaldehyde |
SDS
missing translation for 'documents'