missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Imidazo[1,2-b]pyridazine, 98%, Alfa Aesar™

$56.74 - $252.91

Chemical Identifiers

CAS 766-55-2
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.127
MDL Number MFCD07782103
InChI Key VTVRXITWWZGKHV-UHFFFAOYSA-N
Synonym imidazo 1,2-b pyridazine, imidazo 1,2-b pyridazine, imidazol 1,2-b pyridazine, imdazo1,2-bpyridazine, pubchem11271, acmc-1bczx, imidazo 2,1-f pyridazine, 1,5-diazaindolizine, ksc258e5b, 1,3a,4-triazaindene
PubChem CID 136599
IUPAC Name imidazo[1,2-b]pyridazine
SMILES C1=CC2=NC=CN2N=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6137706 View Documents Alfa Aesar
H6137706
5g Each for $56.74
 
AAH6137714 View Documents Alfa Aesar
H6137714
25g Each for $252.91
 
Description

Description

Applications
Imidazo[1,2-b]pyridazine is used as an intermediate of cefozopran, building block and in fused ring systems.

Solubility
Soluble in dimethylformamide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

766-55-2
119.127
VTVRXITWWZGKHV-UHFFFAOYSA-N
136599
C1=CC2=NC=CN2N=C1
C6H5N3
MFCD07782103
imidazo 1,2-b pyridazine, imidazo 1,2-b pyridazine, imidazol 1,2-b pyridazine, imdazo1,2-bpyridazine, pubchem11271, acmc-1bczx, imidazo 2,1-f pyridazine, 1,5-diazaindolizine, ksc258e5b, 1,3a,4-triazaindene
imidazo[1,2-b]pyridazine

Specifications

54°C
766-55-2
MFCD07782103
VTVRXITWWZGKHV-UHFFFAOYSA-N
imidazo[1,2-b]pyridazine
136599
98%
5g
C6H5N3
imidazo 1,2-b pyridazine, imidazo 1,2-b pyridazine, imidazol 1,2-b pyridazine, imdazo1,2-bpyridazine, pubchem11271, acmc-1bczx, imidazo 2,1-f pyridazine, 1,5-diazaindolizine, ksc258e5b, 1,3a,4-triazaindene
C1=CC2=NC=CN2N=C1
119.127
119.12
Imidazo[1,2-b]pyridazine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger