missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Imidazo[1,2-b]pyridazine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6137714

 View more versions of this product

Catalog No. AAH6137714


Description

Description

Imidazo[1,2-b]pyridazine is used as an intermediate of cefozopran, building block and in fused ring systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Imidazo[1,2-b]pyridazine is used as an intermediate of cefozopran, building block and in fused ring systems.

Solubility
Soluble in dimethylformamide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

Imidazo[1,2-b]pyridazine
54°C
MFCD07782103
imidazo 1,2-b pyridazine, imidazol 1,2-b pyridazine, imdazo1,2-bpyridazine, pubchem11271, acmc-1bczx, imidazo 2,1-f pyridazine, 1,5-diazaindolizine, ksc258e5b, 1,3a,4-triazaindene
VTVRXITWWZGKHV-UHFFFAOYSA-N
imidazo[1,2-b]pyridazine
136599
98%
766-55-2
C6H5N3
25 g
Soluble in dimethylformamide.
C1=CC2=NC=CN2N=C1
119.127
119.12

Chemical Identifiers

766-55-2
119.127
VTVRXITWWZGKHV-UHFFFAOYSA-N
136599
C1=CC2=NC=CN2N=C1
C6H5N3
MFCD07782103
imidazo 1,2-b pyridazine, imidazol 1,2-b pyridazine, imdazo1,2-bpyridazine, pubchem11271, acmc-1bczx, imidazo 2,1-f pyridazine, 1,5-diazaindolizine, ksc258e5b, 1,3a,4-triazaindene
imidazo[1,2-b]pyridazine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only