missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Imidazo[1,2-a]pyrimidine, TCI America™

$212.36 - $740.98

Chemical Identifiers

CAS 274-95-3
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.127
MDL Number MFCD06245377
InChI Key INSWZAQOISIYDT-UHFFFAOYSA-N
PubChem CID 577018
IUPAC Name imidazo[1,2-a]pyrimidine
SMILES C1=CN2C=CN=C2N=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I07981G View Documents TCI America
I07981G
1g Each for $212.36
 
I07985G View Documents TCI America
I07985G
5g Each for $740.98
 
Specifications

Chemical Identifiers

274-95-3
119.127
INSWZAQOISIYDT-UHFFFAOYSA-N
imidazo[1,2-a]pyrimidine
C6H5N3
MFCD06245377
577018
C1=CN2C=CN=C2N=C1

Specifications

Undesignated
1g
C6H5N3
INSWZAQOISIYDT-UHFFFAOYSA-N
imidazo[1,2-a]pyrimidine
577018
Crystal-Powder at 20°C
Imidazo[1,2-a]pyrimidine
127°C
274-95-3
MFCD06245377
C1=CN2C=CN=C2N=C1
119.127
119.13
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No