missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Imidazo[1,2-a]pyridine-8-carboxylic acid hydrochloride, 97%, ACROS Organics™

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C8H6N2O2·ClH
Formula Weight 198.61
Chemical Name or Material Imidazo[1, 2-a]pyridine-8-carboxylic acid hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC440660010 SDS Thermo Fisher Scientific
AC440660010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC440660050 Thermo Fisher Scientific
AC440660050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
C8H6N2O2·ClH
198.61
Imidazo[1, 2-a]pyridine-8-carboxylic acid hydrochloride

Specifications

Powder
1g
Authentic
97%
Glass bottle
Imidazo[1, 2-a]pyridine-8-carboxylic acid hydrochloride
198.61
White to off-white
145335-90-6
C8H6N2O2·ClH
46.44 to 50.32% C, 13.54 to 14.67% N

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.