Learn More
Imidazo[1,2-a]pyridine-8-carboxylic acid hydrochloride, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 440660010
Specifications
Imidazo[1, 2-a]pyridine-8-carboxylic acid hydrochloride | |
46.44 to 50.32% C, 13.54 to 14.67% N | |
97% | |
C8H6N2O2·ClH | |
imidazo 1,2-a pyridine-8-carboxylic acid hydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid monohydrochloride, imidazo 1,2-a pyridine-8-carboxylic acid, monohydrochloride, imidazo 1,2-a pyridine-8-carboxylic acid monohydrochloride, imidazo 1,2-a pyridine-8-carboxylicacidhydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid hydrochloride, imidazo 1,2-alpha pyridine-8-carboxylic acid hydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid monohydr | |
C1=CN2C=CN=C2C(=C1)C(=O)O.Cl | |
198.61 | |
198.61 |
145335-90-6 | |
Authentic | |
Glass bottle | |
1g | |
XRQZBGADESKIBB-UHFFFAOYSA-N | |
imidazo[1,2-a]pyridine-8-carboxylic acid;hydrochloride | |
11715356 | |
97% |
Chemical Identifiers
145335-90-6 | |
198.61 | |
imidazo 1,2-a pyridine-8-carboxylic acid hydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid monohydrochloride, imidazo 1,2-a pyridine-8-carboxylic acid, monohydrochloride, imidazo 1,2-a pyridine-8-carboxylic acid monohydrochloride, imidazo 1,2-a pyridine-8-carboxylicacidhydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid hydrochloride, imidazo 1,2-alpha pyridine-8-carboxylic acid hydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid monohydr | |
imidazo[1,2-a]pyridine-8-carboxylic acid;hydrochloride |
C8H6N2O2·ClH | |
XRQZBGADESKIBB-UHFFFAOYSA-N | |
11715356 | |
C1=CN2C=CN=C2C(=C1)C(=O)O.Cl |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO â Research Use Only