missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Imidazo[1,2-a]pyridine-8-carboxylic acid hydrochloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC440660050

 View more versions of this product

Catalog No. AC440660050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Imidazo[1, 2-a]pyridine-8-carboxylic acid hydrochloride
Authentic
White
5g
97%
imidazo 1,2-a pyridine-8-carboxylic acid hydrochloride, imidazo 1,2-a pyridine-8-carboxylic acid monohydrochloride, imidazo 1,2-a pyridine-8-carboxylic acid, monohydrochloride, imidazo 1,2-a pyridine-8-carboxylicacidhydrochloride, imidazo 1,2-alpha pyridine-8-carboxylic acid hydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid hydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid monohydr, imidazol 1,2-a pyridine-8-carboxylic acid monohydrochloride
C1=CN2C=CN=C2C(=C1)C(=O)O.Cl
198.61
198.61
97%
46.44 to 50.32% C, 13.54 to 14.67% N
Glass bottle
298°C to 300.1°C
145335-90-6
C8H6N2O2·ClH
XRQZBGADESKIBB-UHFFFAOYSA-N
imidazo[1,2-a]pyridine-8-carboxylic acid;hydrochloride
11715356
Powder

Chemical Identifiers

145335-90-6
198.61
imidazo 1,2-a pyridine-8-carboxylic acid hydrochloride, imidazo 1,2-a pyridine-8-carboxylic acid monohydrochloride, imidazo 1,2-a pyridine-8-carboxylic acid, monohydrochloride, imidazo 1,2-a pyridine-8-carboxylicacidhydrochloride, imidazo 1,2-alpha pyridine-8-carboxylic acid hydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid hydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid monohydr, imidazol 1,2-a pyridine-8-carboxylic acid monohydrochloride
imidazo[1,2-a]pyridine-8-carboxylic acid;hydrochloride
C8H6N2O2·ClH
XRQZBGADESKIBB-UHFFFAOYSA-N
11715356
C1=CN2C=CN=C2C(=C1)C(=O)O.Cl
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents