missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Imidazo[1,2-a]pyridine-6-carboxylic acid, 90%, Thermo Scientific™
Chemical Identifiers
CAS | 139022-25-6 |
---|---|
Molecular Formula | C8H6N2O2 |
Molecular Weight (g/mol) | 162.15 |
MDL Number | MFCD07021498 |
InChI Key | ONOJJCTXSDBVSP-UHFFFAOYSA-N |
Synonym | imidazo 1,2-a pyridine-6-carboxylic acid, h-imidazo 1,2-a pyridine-6-carboxylic acid, imidazo 1,2-a pyridine-6-carboxylicacid, 1h-imidazo 1,2-a pyridine-6-carboxylic acid, 4-hydroimidazo 1,2-a pyridine-6-carboxylic acid, pubchem23163, acmc-209cig, ksc173s9l, 6-carboxyimidazo 1,2-a pyridine, imidazol 1,2-a pyridine-6-carboxylic acid |
PubChem CID | 7075376 |
IUPAC Name | imidazo[1,2-a]pyridine-6-carboxylic acid |
SMILES | OC(=O)C1=CN2C=CN=C2C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
CC33101CB
|
Thermo Scientific Chemicals
CC33101CB |
250mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC33101DA
|
Thermo Scientific Chemicals
CC33101DA |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC33101DE
|
Thermo Scientific Chemicals
CC33101DE |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
139022-25-6 | |
162.15 | |
ONOJJCTXSDBVSP-UHFFFAOYSA-N | |
7075376 | |
OC(=O)C1=CN2C=CN=C2C=C1 |
C8H6N2O2 | |
MFCD07021498 | |
imidazo 1,2-a pyridine-6-carboxylic acid, h-imidazo 1,2-a pyridine-6-carboxylic acid, imidazo 1,2-a pyridine-6-carboxylicacid, 1h-imidazo 1,2-a pyridine-6-carboxylic acid, 4-hydroimidazo 1,2-a pyridine-6-carboxylic acid, pubchem23163, acmc-209cig, ksc173s9l, 6-carboxyimidazo 1,2-a pyridine, imidazol 1,2-a pyridine-6-carboxylic acid | |
imidazo[1,2-a]pyridine-6-carboxylic acid |
Specifications
139022-25-6 | |
C8H6N2O2 | |
250mg | |
ONOJJCTXSDBVSP-UHFFFAOYSA-N | |
imidazo[1,2-a]pyridine-6-carboxylic acid | |
7075376 | |
97% |
97% | |
MFCD07021498 | |
imidazo 1,2-a pyridine-6-carboxylic acid, h-imidazo 1,2-a pyridine-6-carboxylic acid, imidazo 1,2-a pyridine-6-carboxylicacid, 1h-imidazo 1,2-a pyridine-6-carboxylic acid, 4-hydroimidazo 1,2-a pyridine-6-carboxylic acid, pubchem23163, acmc-209cig, ksc173s9l, 6-carboxyimidazo 1,2-a pyridine, imidazol 1,2-a pyridine-6-carboxylic acid | |
OC(=O)C1=CN2C=CN=C2C=C1 | |
162.15 | |
162.15 | |
Imidazo[1, 2-a]pyridine-6-carboxylic acid |
Safety and Handling
Warning