Please login to your online account to display your discounted pricing

Imidazo[1,2-a]pyridine-6-carboxylic acid, 90%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  CC33101CB

 View more versions of this product

Catalog No. CC33101CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 162.15
Quantity 250mg
Assay 97%
Chemical Name or Material Imidazo[1, 2-a]pyridine-6-carboxylic acid
Assay Percent Range 97%
Molecular Formula C8H6N2O2
CAS 139022-25-6
Health Hazard 1 Warning

Specifications

Chemical Name or Material Imidazo[1, 2-a]pyridine-6-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H6N2O2
Formula Weight 162.15
Accessories

Warning