missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Imidazo[1,2-a]pyridine-6-carbaldehyde, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC33104DA

 View more versions of this product

Catalog No. CC33104DA



Description

Description

Specifications

Specifications

Imidazo[1, 2-a]pyridine-6-carbaldehyde
94.5
95%
C8H6N2O
1 g
VDWYLKPRUHCLAW-UHFFFAOYSA-N
imidazo[1,2-a]pyridine-6-carbaldehyde
7164582
95%
116355-16-9
100.0
Amber Glass Bottle
MFCD07772808
imidazo 1,2-a pyridine-6-carbaldehyde, imidazo 1,2-a pyridine-6-carboxaldehyde, 6-formylimidazo 1,2-a pyridine, pubchem23883, acmc-20a31e, 6-imidazo 1,2-a pyridinecarbaldehyde, 6-imidazo 1,2-a-pyridinecarbaldehyde, h-imidazo 1,2-a pyridine-6-carbaldehyde, 4-hydroimidazo 1,2-a pyridine-6-carbaldehyde
C1=CC2=NC=CN2C=C1C=O
146.149
146.15

Chemical Identifiers

116355-16-9
146.149
VDWYLKPRUHCLAW-UHFFFAOYSA-N
7164582
C1=CC2=NC=CN2C=C1C=O
C8H6N2O
MFCD07772808
imidazo 1,2-a pyridine-6-carbaldehyde, imidazo 1,2-a pyridine-6-carboxaldehyde, 6-formylimidazo 1,2-a pyridine, pubchem23883, acmc-20a31e, 6-imidazo 1,2-a pyridinecarbaldehyde, 6-imidazo 1,2-a-pyridinecarbaldehyde, h-imidazo 1,2-a pyridine-6-carbaldehyde, 4-hydroimidazo 1,2-a pyridine-6-carbaldehyde
imidazo[1,2-a]pyridine-6-carbaldehyde
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions