missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Imidazo[1,2-a]pyridine-2-carbaldehyde, ≥97%, Maybridge

Chemical Identifiers

CAS 118000-43-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H6N2O
Formula Weight 146.15
MDL Number MFCD06739247
Synonym imidazo(1, 2-a)pyridine-2-carbaldehyde (IUPAC)
Chemical Name or Material Imidazo[1, 2-a]pyridine-2-carbaldehyde
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC31304CB SDS Fisher Scientific Company
CC31304CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC31304DA SDS Fisher Scientific Company
CC31304DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC31304DE Fisher Scientific Company
CC31304DE
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

118000-43-4
100.0
C8H6N2O
MFCD06739247
Imidazo[1, 2-a]pyridine-2-carbaldehyde
96.5
97%
146.15
imidazo(1, 2-a)pyridine-2-carbaldehyde (IUPAC)
97%

Specifications

146.15
100.0
C8H6N2O
imidazo(1, 2-a)pyridine-2-carbaldehyde (IUPAC)
97%
Imidazo[1, 2-a]pyridine-2-carbaldehyde
250mg
118000-43-4
96.5
Amber Glass Bottle
MFCD06739247
97%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.