Please login to your online account to display your discounted pricing

Imidazo[1,2-a]pyridine-2-carbaldehyde, ≥97%, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  CC31304DE

 View more versions of this product

Catalog No. CC31304DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 146.15
Quantity 5g
Assay 97%
CAS Max % 100.0
Synonym imidazo(1, 2-a)pyridine-2-carbaldehyde (IUPAC)
Molecular Formula C8H6N2O
CAS 118000-43-4
Assay Percent Range 97%
CAS Min % 96.5
MDL Number MFCD06739247
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Imidazo[1, 2-a]pyridine-2-carbaldehyde
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Imidazo[1, 2-a]pyridine-2-carbaldehyde
CAS 118000-43-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H6N2O
Formula Weight 146.15
MDL Number MFCD06739247
Synonym imidazo(1, 2-a)pyridine-2-carbaldehyde (IUPAC)
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.