Please login to your online account to display your discounted pricing

Imidazo[1,2-a]pyridin-6-ylmethanol, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  CC33109CB

 View more versions of this product

Catalog No. CC33109CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 148.16
Quantity 250mg
Assay 97%
CAS 132213-07-1
Chemical Name or Material Imidazo[1, 2-a]pyridin-6-ylmethanol
Assay Percent Range 97%
Molecular Formula C8H8N2O

Specifications

Chemical Name or Material Imidazo[1, 2-a]pyridin-6-ylmethanol
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H8N2O
Formula Weight 148.16