Please login to your online account to display your discounted pricing

iMAC2, Tocris™

Suppressor of mitochondrial apoptosis

$259.29 - $1,075.94

Chemical Identifiers

Chemical Name or Material iMAC2
Assay Percent Range >98%
Molecular Formula C19H20Br2FN3·2HCl
Formula Weight 542.11
Synonym 3, 6-Dibromo-9-[2-fluoro-3-(1-piperazinyl)propyl]-9H-carbazole dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

379410

 
r & d systems
379410
10mg Each for $259.29

379450

 
r & d systems
379450
50mg Each for $1,075.94
Description & Specifications

Specifications

Formula Weight 542.11
Quantity 10mg
Chemical Name or Material iMAC2
CAS 335166-00-2
Synonym 3, 6-Dibromo-9-[2-fluoro-3-(1-piperazinyl)propyl]-9H-carbazole dihydrochloride
Recommended Storage Desiccate at Room Temperature
Assay Percent Range >98%
Molecular Formula C19H20Br2FN3·2HCl
Safety and Handling

Desiccate at Room Temperature