Fisher Scientific Illuminated Fingerprint Magnifying Glass
Slips in a shirt or jacket pocket — always ready when you need it.
Supplier:  Fisher Scientific 314M
Catalog No. S99186
Requires : Two AA penlight batteries (S43921, not included)
Description
Description
The go-anywhere tool for locating evidence at the crime scene.
  • Built-in illuminator
  • Powerful 4X lens
  • Housed in a tough, lightweight, plastic case
Specifications
Specifications
Magnifier, Flashlight
Illumination Lamp
4x
For locating evidence at the crime scene, for classifying fingerprints and examining evidence
Glass, Plastic