Please login to your online account to display your discounted pricing

Alfa Aesar™ Ilimaquinone, 98%

Chemical Identifiers

Chemical Name or Material Ilimaquinone
CAS 71678-03-0
Formula Weight 358.48
MDL Number MFCD22200891
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67278EXG

 
alfa aesar™
J67278EXG
100μg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 358.48
Chemical Name or Material Ilimaquinone
Physical Form Oil
Assay 98%
MDL Number MFCD22200891
CAS 71678-03-0
Color Yellow
Quantity 100μg
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C