Please login to your online account to display your discounted pricing

ILC Dover Headcover

$681.26 - $681.26

Specifications

Color White
Hood Material Polyethylene; Polyolefin
Material Polyethylene/Polyolefin
Size Universal
For Use With (Equipment) Sentinel XL HP system, CBRN system
View More Specs
Products
Catalog Number Mfr. No. Includes Price Quantity    

19-631-045

 
ilc dover llc
DOVS400110
Ten head covers Pack of 10 Each for $681.26
Description & Specifications

Specifications

Color White
Hood Material Polyethylene; Polyolefin
Material Polyethylene/Polyolefin
Size Universal
For Use With (Equipment) Sentinel XL HP system, CBRN system
Lens Material PET; Polyester Film