Please login to your online account to display your discounted pricing

ILC Dover Colored Headcover

$1,118.05 - $1,118.05

Specifications

Color Multi-colored
Hood Material Polyethylene; Polyolefin
Size Universal
For Use With (Equipment) Sentinel XL HP system, CBRN system
Lens Material PET
View More Specs
Products
Catalog Number Mfr. No. Includes Price Quantity    

19-631-048

 
ilc dover llc
DOVS4001P10
Ten BioShield head covers Each for $1,118.05
Description & Specifications

Specifications

Color Multi-colored
Hood Material Polyethylene; Polyolefin
Size Universal
For Use With (Equipment) Sentinel XL HP system, CBRN system
Lens Material PET