Please login to your online account to display your discounted pricing

IL-9 Mouse anti-Human, PerCP-eFluor 710, Clone: MH9D1, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $404.55

Specifications

Antigen IL-9
Clone MH9D1
Host Species Mouse
Gene Alias Interleukin-9, IL9, T-cell growth factor P40
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123891

 
life technologies
LS46709841
25 tests Each for $172.55

501123892

 
life technologies
LS46709842
100 tests Each for $404.55
Description & Specifications

Specifications

Antigen IL-9
Clone MH9D1
Host Species Mouse
Gene Alias Interleukin-9, IL9, T-cell growth factor P40
Species Reactivity Human
Applications Flow Cytometry
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.03μg)/test

The monoclonal antibody MH9D1 recognizes interleukin-9 (IL-9), a proinflammatory cytokine historically believed to be involved in type 2 immune responses. Recent evidence suggests IL-9 may instead be secreted by a distinct T-helper lineage called Th9. These cells can be derived from Th2 cells with TGF beta or differentiated directly from naïve CD4+ T cells with TGF beta and IL-4.

IL-9 is a member of the common cytokine receptor gamma chain-dependent family of cytokines which also includes IL-2, IL-4, IL-7, IL-15 and IL-21. Its pleiotropic effects on Th2 lymphocytes, B lymphocytes, mast cells, eosinophils and gut and airway epithelial cells have implicated IL-9 in asthma and other allergy-related disorders. Staining with the MH9D1 antibody is not blocked by the MH9A4 antibody, suggesting that the two antibodies bind to unique epitopes.