Learn More
IL-9 Monoclonal Antibody (MH9A4), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$197.00 - $197.00
Specifications
Antigen | IL-9 |
---|---|
Clone | MH9A4 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | eFluor 660 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
501124433
|
Invitrogen
50709741 |
25 Tests |
Each for $197.00
|
|
|||||
Description
Interleukin 9 (IL-9) is produced by activated T helper 2 (Th2) lymphocytes. IL-9 regulates many cell types involved in classical Th2-associated asthma responses, including B and T lymphocytes, mast cells, eosinophils, and epithelial cells. The mouse IL-9 cDNA encodes a 144 amino acid precursor protein with an 18 amino acid signal peptide that is cleaved to form the mature cysteine-rich protein with a molecular mass of 14 kDa. Native mouse IL-9 is heavily glycosylated. Recently, TGF beta has been reported to play an important role in the differentiation of IL-17-producing T helper cells and inducible regulatory T cells by reprogramming Th2 cells to secrete IL-9, thereby driving the differentiation of Th9 cells directly.
IL-9 Monoclonal antibody specifically detects IL-9 in Human samples. It is validated for Flow Cytometry.Specifications
IL-9 | |
Flow Cytometry | |
eFluor 660 | |
Mouse | |
Human | |
3578 | |
Primary | |
Antibody |
MH9A4 | |
Monoclonal | |
Liquid | |
RUO | |
P15248 | |
IgG2b κ | |
Affinity chromatography |
For Research Use Only.