Please login to your online account to display your discounted pricing

IL-9 Mouse anti-Human, eFluor 450, Clone: MH9D1, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $433.55

Specifications

Antigen IL-9
Clone MH9D1
Host Species Mouse
Gene Alias Interleukin-9
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124201

 
life technologies
LS48709841
25 tests Each for $172.55

501124202

 
life technologies
LS48709842
100 tests Each for $433.55
Description & Specifications

Specifications

Antigen IL-9
Clone MH9D1
Host Species Mouse
Gene Alias Interleukin-9
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The monoclonal antibody MH9D1 recognizes interleukin-9 (IL-9), a proinflammatory cytokine historically believed to be involved in type 2 immune responses. Recent evidence suggests IL-9 may instead be secreted by a distinct T-helper lineage called Th9. These cells can be derived from Th2 cells with TGF beta or differentiated directly from naïve CD4+ T cells with TGF beta and IL-4.

IL-9 is a member of the common cytokine receptor gamma chain-dependent family of cytokines which also includes IL-2, IL-4, IL-7, IL-15 and IL-21. Its pleiotropic effects on Th2 lymphocytes, B lymphocytes, mast cells, eosinophils and gut and airway epithelial cells have implicated IL-9 in asthma and other allergy-related disorders. Staining with the MH9D1 antibody is not blocked by the MH9A4 antibody, suggesting that the two antibodies bind to unique epitopes.