missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-6 Rat anti-Mouse, PE, Clone: MP5-20F3, eBioscience™

Rat Monoclonal Antibody

$130.68 - $289.08

Specifications

Antigen IL-6
Clone MP5-20F3
Host Species Rat
Gene Alias Interleukin-6
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011135 Life Technologies
LS12706141
25 Tests Each for $130.68
5011136 Life Technologies
LS12706181
50μg Each for $157.08
5011137 Life Technologies
LS12706182
100μg Each for $289.08
Specifications

Description

The MP5-20F3 antibody reacts with mouse interleukin-6 (IL-6). The MP5-20F3 antibody is a neutralizing antibody. Mouse IL-6 (also known as B Cell Stimulating Factor, BCSF, and BSF-2) is a 21-28kDa factor produced by a variety of cell types, including monocytes, fibroblasts and endothelial cells. Upon stimulation, macrophages, T, B, mast, and glial cells, eosinophils, keratinocytes and granulocytes also secrete IL-6. This pleiotrophic cytokine influences the antigen-specific immune response, inflammatory response, and the development of neurons, and is a major mediator of the acute phase reaction.
Description & Specifications

Specifications

IL-6
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
MP5-20F3
Interleukin-6
Flow Cytometry (Intracellular Staining)
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL; 5μL (0.125μg)/test