Please login to your online account to display your discounted pricing

IL-6 Rat anti-Mouse, Biotin, Clone: MP5-32C11, eBioscience™

Rat Monoclonal Antibody

$143.55 - $375.55

Specifications

Antigen IL-6
Clone MP5-32C11
Host Species Rat
Gene Alias Interleukin-6, IL6
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012163

 
life technologies
LS13706281
50μg Each for $143.55

5012164

 
life technologies
LS13706285
500μg Each for $375.55
Description & Specifications

Specifications

Antigen IL-6
Clone MP5-32C11
Host Species Rat
Gene Alias Interleukin-6, IL6
Species Reactivity Mouse
Applications ELISA Detection
Applications ELISPOT Detection Antibody
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The MP5-32C11 antibody reacts with mouse interleukin-6 (IL-6), a 21-28kDa cytokine secreted by a variety of cell types.