Please login to your online account to display your discounted pricing

IL-6 Rat anti-Human, PE-Cy7, Clone: MQ2-13A5, eBioscience™

Rat Monoclonal Antibody

$201.55 - $477.05

Specifications

Antigen IL-6
Clone MQ2-13A5
Host Species Rat
Gene Alias Interleukin-6
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123432

 
life technologies
LS25706941
25 tests Each for $201.55

501123433

 
life technologies
LS25706942
100 tests Each for $477.05
Description & Specifications

Specifications

Antigen IL-6
Clone MQ2-13A5
Host Species Rat
Gene Alias Interleukin-6
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

The MQ2-13A5 antibody reacts with human interleukin-6 (IL-6), a 21-28kDa cytokine secreted by a variety of cell types.