Please login to your online account to display your discounted pricing

IL-6 Rat anti-Human, PE, Clone: MQ2-13A5, eBioscience™

Rat Monoclonal Antibody

$143.55 - $375.55

Specifications

Antigen IL-6
Clone MQ2-13A5
Host Species Rat
Gene Alias Interleukin-6, IL6
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011138

 
life technologies
LS12706941
25 tests Each for $143.55

5011139

 
life technologies
LS12706981
50μg Each for $201.55

5011140

 
life technologies
LS12706982
100μg Each for $375.55
Description & Specifications

Specifications

Antigen IL-6
Clone MQ2-13A5
Host Species Rat
Gene Alias Interleukin-6, IL6
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL; 5μL (0.125μg)/test

The MQ2-13A5 antibody reacts with human interleukin-6 (IL-6), a 21-28kDa cytokine secreted by a variety of cell types.