Please login to your online account to display your discounted pricing

IL-6 Rat anti-Human, Clone: MQ2-39C3, eBioscience™

Rat Monoclonal Antibody

$114.55 - $332.05

Specifications

Antigen IL-6
Clone MQ2-39C3
Host Species Rat
Gene Alias Interleukin-6, IL6
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013616

 
life technologies
LS14706881
50μg Each for $114.55

5013617

 
life technologies
LS14706885
500μg Each for $332.05
Description & Specifications

Specifications

Antigen IL-6
Clone MQ2-39C3
Host Species Rat
Gene Alias Interleukin-6, IL6
Species Reactivity Human
Applications ELISA
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The MQ2-39C3 antibody reacts with human interleukin-6 (IL-6), a 21-28kDa cytokine secreted by a variety of cell types.