missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-6 Rat anti-Human, Clone: MQ2-13A5, Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$130.68 - $2,242.68

Specifications

Antigen IL-6
Clone MQ2-13A5
Host Species Rat
Gene Alias Interleukin-6
Applications Cytokine Neutralization,ELISA Capture,ELISPOT
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122982 Life Technologies
LS16706938
5mg Each for $2,242.68
5014706 Life Technologies
LS16706981
50μg Each for $130.68
5014707 Life Technologies
LS16706985
500μg Each for $381.48
Specifications

Description

The MQ2-13A5 antibody reacts with human interleukin-6 (IL-6), a 21-28kDa cytokine secreted by a variety of cell types.
Description & Specifications

Specifications

IL-6
Rat
Cytokine Neutralization,ELISA Capture,ELISPOT
Unlabeled
Store at 2-8°C.
Monoclonal
1mg/mL
MQ2-13A5
Interleukin-6
RUO
Purified
Primary
aqueous buffer, no sodium azide